Assay Details
| Gene Symbol: Rhov | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_145530.2 | NP_663505.1 | 2 - 3 | 368 | 71 | |
| GenBank mRNA | AK036775.1 | - | 2 - 3 | 369 | 71 | |
| AK037170.1 | - | 2 - 3 | 370 | 71 | ||
| BC021307.1 | - | 2 - 3 | 373 | 5003249 | 71 | |
More Information
Updated Mapping Information
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P21568741 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P21568739 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P21568738 | - | - |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P439584 | AK037170.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P439584 | AK036775.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P439584 | NM_145530.2 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P439584 | AK037170.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P439584 | AK036775.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P439584 | NM_145530.2 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 624943 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P123577 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P439584 | AK037170.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P439584 | AK036775.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P439584 | NM_145530.2 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P439584 | AK037170.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P439584 | AK036775.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P439584 | NM_145530.2 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P439584 | AK037170.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P439584 | AK036775.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P439584 | NM_145530.2 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P439584 | AK037170.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P439584 | AK036775.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P439584 | NM_145530.2 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21568739 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21568737 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21568743 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21568742 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21568723 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21568732 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21568754 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21568752 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21568736 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21568734 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21568725 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21568745 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21568727 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21568755 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21568749 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21568756 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21568738 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21568730 | - | - |
| Whole Mouse Genome G4122A | A_51_P439584 | AK037170.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P439584 | AK036775.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P439584 | NM_145530.2 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P439584 | AK037170.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P439584 | AK036775.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P439584 | NM_145530.2 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P129384 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P02083738 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P02085488 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P02085469 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P21568738 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P21568739 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P21568741 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21568721 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21568673 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21568670 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21568710 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21568776 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21568764 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21568732 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21568709 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21568736 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21568676 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21568716 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21568684 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21568761 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21568738 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21568715 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21568694 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21568717 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21568713 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21568739 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21568743 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21568754 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21568727 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21568702 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21568685 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21568692 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21568690 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21568686 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21568701 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21568703 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21568747 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21568725 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21568683 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21568699 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21568697 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21568737 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21568769 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21568753 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21568742 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21568734 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21568758 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21568714 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21568707 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21568770 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21568705 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21568730 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21568766 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21568756 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21568750 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21568723 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01824097 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21568686 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21568701 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21568747 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21568720 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21568749 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21568770 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21568754 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21568691 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21568715 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21568676 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21568702 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21568736 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21568674 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21568699 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P02085469 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21568700 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01447971 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21568712 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21568685 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21568761 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01522810 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21568711 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21568734 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01634482 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21568709 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21568684 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21568756 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01257789 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P02280220 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21568695 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21568696 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P02085483 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21568746 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21568737 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21568745 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P02085493 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21568682 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01577164 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21568744 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21568732 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01274903 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01577166 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21568766 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21568688 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21568753 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21568705 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P02239221 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21568769 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01560469 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21568714 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21568717 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01634483 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21568727 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21568742 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21568707 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21568772 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P02085488 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21568730 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10486198 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10486199 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10486200 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1424976_PM_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10486197 | - | - |
| Mouse Genome 430 2.0 Array | 1424976_at | - | - |
| Mouse Genome 430 2.0 Array | 1424976_PM_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 228543_PM_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 228543_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR0200036985.mm.1 | NM_145530.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0200036985.mm.1 | NM_145530.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0200036985.mm.1 | TC0200004427.mm.1 | MTA TC |
| Mouse Expression Set 430 | 1424976_at | - | - |
| Mouse Expression Set 430 | 1424976_PM_at | - | - |
| Murine Genome U74v2 Set | 133045_PM_at | - | - |
| Murine Genome U74v2 Set | 133045_at | - | - |
| Mouse Genome 430A 2.0 Array | 1424976_at | - | - |
| Mouse Genome 430A 2.0 Array | 1424976_PM_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1424976_PM_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1424976_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 850496 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina mouse-6 v1.0 expression beadchip | 5860100 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0005860100 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_1244918 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_1244918 | - | - |
| mouse-6 v1.1 (Illumina) | scl18807.3_31-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 5860100 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl18807.3_31-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 5860100 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_2859528 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2859528 | - | - |

Rhov
Genomic Map