Assay Details
| Gene Symbol: Nostrin | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_181547.3 | NP_853525.3 | 10 - 11 | 949 | 99 | |
| GenBank mRNA | AJ564264.1 | - | 10 - 11 | 859 | 99 | |
| AY280963.1 | - | 10 - 11 | 902 | 99 | ||
| BC069942.1 | - | 10 - 11 | 926 | 30443942 | 99 | |
More Information
Updated Mapping Information
Set Membership: |
|

Nostrin
Genomic Map