Assay Details
| Gene Symbol: Nostrin | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_181547.3 | NP_853525.3 | 3 - 4 | 291 | 79 | |
| GenBank mRNA | AJ564264.1 | - | 3 - 4 | 201 | 79 | |
| AK052926.1 | - | 3 - 4 | 283 | 79 | ||
| AY280963.1 | - | 3 - 4 | 244 | 79 | ||
| BC069942.1 | - | 3 - 4 | 268 | 30443942 | 79 | |
| BY061165.1 | - | 3 - 4 | 291 | 79 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Nostrin
Genomic Map