Assay Details
| Gene Symbol: Gpr68 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001177673.1 | NP_001171144.1 | 1 - 2 | 107 | 133 | |
| NM_175493.4 | NP_780702.1 | 1 - 2 | 107 | 133 | ||
| GenBank mRNA | AK087410.1 | - | 1 - 2 | 91 | 133 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the AK087410.1, NM_001177673.1, NM_175493.4 transcripts.
Set Membership: |
|

Gpr68
Genomic Map