Assay Details
| Gene Symbol: Cdyl | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001123386.1 | NP_001116858.1 | 5 - 6 | 1794 | 68 | |
| NM_009881.3 | NP_034011.1 | 8 - 9 | 1744 | 68 | ||
| GenBank mRNA | AF081260.1 | - | 8 - 9 | 1726 | 68 | |
| AF081261.1 | - | 8 - 9 | 1726 | 68 | ||
| AK028162.1 | - | 5 - 6 | 1490 | 68 | ||
| AK156509.1 | - | 5 - 6 | 68 | |||
| AK162180.1 | - | 3 - 4 | 474 | 68 | ||
| BC055103.1 | - | 8 - 9 | 1693 | 6741468 | 68 | |
| BC062123.1 | - | 5 - 6 | 1883 | 6826210 | 68 | |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Cdyl
Genomic Map