Assay Details
| Gene Symbol: 4921508D12Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_045802.1 | 1 - 2 | 122 | 80 | ||
| GenBank mRNA | AK014842.1 | - | 1 - 2 | 124 | 80 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | prokineticin receptor 2 | Prokr2 |
B830005M06Rik, EG-VEGRF2, Gpcr73l1, Gpr73l1, PKR2 |
-39.3kb | Anti-sense | Chr. 2 - 132370329 - 132385498 on NCBI Build GRCm38 |
| Downstream | RIKEN cDNA 1700026D11 gene | 1700026D11Rik | 45.1kb | Sense | Chr. 2 - 132490728 - 132524057 on NCBI Build GRCm38 |
More Information
Additional Information:
This assay is in the 5'UTR region of the AK014842.1 transcripts.
Set Membership: |
|

4921508D12Rik
Genomic Map