Assay Details
| Gene Symbol: 4921508D12Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_045802.1 | 2 - 3 | 290 | 66 | ||
| GenBank mRNA | AK014842.1 | - | 2 - 3 | 292 | 66 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | prokineticin receptor 2 | Prokr2 |
B830005M06Rik, EG-VEGRF2, Gpcr73l1, Gpr73l1, PKR2 |
-60.0kb | Anti-sense | Chr. 2 - 132370329 - 132385498 on NCBI Build GRCm38 |
| Downstream | RIKEN cDNA 1700026D11 gene | 1700026D11Rik | 42.7kb | Sense | Chr. 2 - 132490728 - 132524057 on NCBI Build GRCm38 |
More Information
Additional Information:
This assay is in the 5'UTR region of the AK014842.1 transcripts.
Set Membership: |
|
MicroArray Probes:
Agilent
Affymetrix
ABI
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P383884 | AK014842.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P383884 | AK014842.1 | mouse_transcriptome |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P132539 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P167348 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P383884 | AK014842.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P383884 | AK014842.1 | mouse_transcriptome |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P132539 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P167348 | - | - |
| Whole Mouse Genome G4122A | A_51_P383884 | AK014842.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P383884 | AK014842.1 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P135899 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P167348 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P132539 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21640386 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21640371 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21640344 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21640362 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21640202 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21640340 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21640345 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21640284 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21640342 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01791482 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21640349 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21640251 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21640242 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21640373 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01448772 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01165022 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P02085787 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P02189110 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P02189115 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P02131072 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21640337 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21640365 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21640401 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21640402 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21640339 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21640390 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01379022 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21640465 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21640276 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01165004 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21640351 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21640347 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21640279 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21640273 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01455551 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21640358 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21640352 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01360689 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21640245 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21640283 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21640263 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P02097248 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21640396 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21640462 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01837159 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21640266 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21640469 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01360682 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01455555 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21640354 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21640389 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P02058292 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21640466 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21640375 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21640265 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21640391 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01837160 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21640333 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21640274 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21640395 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21640241 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21640275 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21640268 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21640460 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21640201 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix HT MG-430 PM Array Plate | 1454188_PM_at | - | - |
| Mouse Genome 430 2.0 Array | 1454188_PM_at | - | - |
| Mouse Genome 430 2.0 Array | 1454188_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR0200016186.mm.1 | NR_045802.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0200016186.mm.1 | NR_045802.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0200016186.mm.1 | TC0200002033.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0200016187.mm.1 | NR_045802.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0200016187.mm.1 | NR_045802.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0200016187.mm.1 | TC0200002033.mm.1 | MTA TC |
| Mouse Expression Set 430 | 1454188_PM_at | - | - |
| Mouse Expression Set 430 | 1454188_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1454188_PM_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1454188_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 354124 | - | - |

4921508D12Rik
Genomic Map