Assay Details
| Gene Symbol: Nox3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_198958.2 | NP_945196.2 | 5 - 6 | 550 | 81 | |
| GenBank mRNA | AY182377.1 | - | 5 - 6 | 550 | 81 | |
| AY573240.1 | - | 5 - 6 | 81 | |||
| BC106862.2 | - | 5 - 6 | 550 | 40047310 | 81 | |
More Information
Updated Mapping Information
Set Membership: |
|

Nox3
Genomic Map