Assay Details
| Gene Symbol: Nox3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_198958.2 | NP_945196.2 | 11 - 12 | 1520 | 103 | |
| GenBank mRNA | AY182377.1 | - | 11 - 12 | 1520 | 103 | |
| AY573240.1 | - | 11 - 12 | 1460 | 103 | ||
| BC106862.2 | - | 11 - 12 | 1520 | 40047310 | 103 | |
More Information
Updated Mapping Information
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P412457 | BC106862.2 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P412457 | NM_198958.2 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P412457 | AY182377.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P412457 | AY573240.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P412457 | BC106862.2 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P412457 | NM_198958.2 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P412457 | AY182377.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P412457 | AY573240.1 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 205050 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 933301 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 453505 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 877813 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 543483 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 141393 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 579825 | AK002989.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 543855 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 530622 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 650858 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 532701 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 681020 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 952792 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 584857 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 368950 | AJ277114.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 368950 | AK131598.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 368950 | AK138441.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 368950 | AK169006.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 368950 | AK179193.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 368950 | BC023319.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 368950 | BC058581.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 368950 | NM_030700.1 | refseq_mrna_mouse_NM |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 278341 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 384765 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 886532 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 873491 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 398671 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 579672 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 189217 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 687782 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 97409 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 928712 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 76935 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 216838 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 476541 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 216958 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 597253 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 616237 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 394116 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 319046 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 88631 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 772302 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 899644 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 886218 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 867064 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 127482 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 8411 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 93217 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 141191 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 86072 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 155424 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 63409 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P139720 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P179668 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P412457 | BC106862.2 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P412457 | NM_198958.2 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P412457 | AY182377.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P412457 | AY573240.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P412457 | BC106862.2 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P412457 | NM_198958.2 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P412457 | AY182377.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P412457 | AY573240.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_52_P412457 | BC106862.2 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_52_P412457 | NM_198958.2 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_52_P412457 | AY182377.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_52_P412457 | AY573240.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_52_P412457 | BC106862.2 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_52_P412457 | NM_198958.2 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_52_P412457 | AY182377.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_52_P412457 | AY573240.1 | mouse_transcriptome |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P03268760 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P08701195 | - | - |
| Whole Mouse Genome G4122A | A_52_P412457 | BC106862.2 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P412457 | NM_198958.2 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P412457 | AY182377.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P412457 | AY573240.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P412457 | BC106862.2 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P412457 | NM_198958.2 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P412457 | AY182377.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P412457 | AY573240.1 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P133123 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_67_P08701195 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P13525020 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P13525028 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P13525014 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P13365694 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P13525026 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31006587 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31006574 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31006523 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31006514 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31006540 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31006538 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31006590 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31006600 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31006534 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31006561 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31006563 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31006537 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31006500 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31006508 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31006521 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31006512 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31006586 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31006592 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31006565 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31006567 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31006589 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31006542 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31006571 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31006545 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31006517 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31006562 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31006569 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31006559 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31006505 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31006530 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31006593 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31006594 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31006580 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31006548 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31006510 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31006539 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31006507 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31006543 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31006554 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31006595 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31006575 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31006515 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31006577 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31006519 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31006532 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31006552 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31006528 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31006598 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31006503 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31006547 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31006550 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31006525 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31006583 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31006509 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31006573 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13206597 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31006554 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31006589 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31006557 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31006546 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31006550 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31006580 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31006586 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31006539 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13206588 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31006579 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31006562 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31006565 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31006568 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31006575 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31006558 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31006537 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31006567 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13155975 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31006569 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31006587 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13155955 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31006590 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31006563 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31006538 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31006561 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31006547 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13206586 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31006574 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31006535 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31006540 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31006552 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31006571 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13155997 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31006577 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31006548 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10447544 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10447533 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10447540 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10447534 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10447532 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10447538 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10447541 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10447531 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10447539 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10447545 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10447535 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10447543 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10447542 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10447536 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10447537 | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10447530 | - | - |
| Affymetrix Mouse Clariom S Array | PSR1700012311.mm.1 | NM_198958.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1700012311.mm.1 | NM_198958.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1700012311.mm.1 | TC1700001358.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1700012310.mm.1 | NM_198958.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1700012310.mm.1 | NM_198958.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1700012310.mm.1 | TC1700001358.mm.1 | MTA TC |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 890130 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| MouseWG-6 v2 Expression BeadChip | 2100358 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 2100358 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0002100358 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2640368 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_2640368 | - | - |
| mouse-6 v1.1 (Illumina) | scl0224480.11_51-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 2100358 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl0224480.11_51-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 2100358 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_2640368 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2640368 | - | - |

Nox3
Genomic Map