Assay Details
| Gene Symbol: Lrch2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001081173.1 | NP_001074642.1 | 14 - 15 | 1747 | 73 | |
| GenBank mRNA | AK122516.1 | - | 3 - 4 | 1562 | 73 | |
| BC056180.1 | - | 8 - 9 | 701 | 73 | ||
More Information
Set Membership: |
|

Lrch2
Genomic Map