Assay Details
| Gene Symbol: Lrch2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001081173.1 | NP_001074642.1 | 3 - 4 | 676 | 92 | |
| GenBank mRNA | AK144916.1 | - | 3 - 4 | 506 | 92 | |
More Information
Updated Mapping Information
Set Membership: |
|

Lrch2
Genomic Map