Assay Details
| Gene Symbol: Sppl2a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_023220.2 | NP_075709.2 | 11 - 12 | 1387 | 86 | |
| GenBank mRNA | AB041547.1 | - | 11 - 12 | 1387 | 86 | |
| AK155011.1 | - | 11 - 12 | 1186 | 86 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Sppl2a
Genomic Map