Assay Details
| Gene Symbol: Sppl2a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_023220.2 | NP_075709.2 | 4 - 5 | 684 | 139 | |
| GenBank mRNA | AB041547.1 | - | 4 - 5 | 684 | 139 | |
| AK155011.1 | - | 4 - 5 | 485 | 139 | ||
| BC026578.1 | - | 4 - 5 | 492 | 5059823 | 139 | |
More Information
Updated Mapping Information
Set Membership: |
|

Sppl2a
Genomic Map