Assay Details
| Gene Symbol: Aspg | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001081169.1 | NP_001074638.1 | 12 - 13 | 1637 | 65 | |
| GenBank mRNA | BC089617.1 | - | 12 - 13 | 1464 | 65 | |
| BC126962.1 | - | 12 - 13 | 1455 | 40092851 | 65 | |
More Information
Updated Mapping Information
Set Membership: |
|

Aspg
Genomic Map