Assay Details
| Gene Symbol: Aspg | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001081169.1 | NP_001074638.1 | 13 - 14 | 1717 | 60 | |
| GenBank mRNA | BC023353.1 | - | 4 - 5 | 502 | 60 | |
| BC089617.1 | - | 13 - 14 | 1544 | 60 | ||
| BC126962.1 | - | 13 - 14 | 1535 | 40092851 | 60 | |
More Information
Updated Mapping Information
Set Membership: |
|

Aspg
Genomic Map