Assay Details
| Gene Symbol: Aspg | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001081169.1 | NP_001074638.1 | 6 - 7 | 846 | 84 | |
| GenBank mRNA | BC089617.1 | - | 6 - 7 | 673 | 84 | |
| BC126962.1 | - | 6 - 7 | 664 | 40092851 | 84 | |
More Information
Updated Mapping Information
Set Membership: |
|

Aspg
Genomic Map