Assay Details
| Gene Symbol: Aspg | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001081169.1 | NP_001074638.1 | 8 - 9 | 1145 | 64 | |
| GenBank mRNA | BC126962.1 | - | 8 - 9 | 963 | 40092851 | 64 |
More Information
Set Membership: |
|

Aspg
Genomic Map