Assay Details
| Gene Symbol: Arpc5l | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_028809.1 | NP_083085.1 | 3 - 4 | 461 | 158 | |
| GenBank mRNA | AK008255.1 | - | 3 - 4 | 461 | 158 | |
| AK037305.1 | - | 2 - 3 | 3352 | 158 | ||
| BC024482.1 | - | 3 - 4 | 484 | 4977391 | 158 | |
More Information
Updated Mapping Information
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P21151651 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P21151650 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P21151655 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P21151656 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P21151652 | - | - |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2067226 | BC024482.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2067226 | NM_028809.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2067226 | AK008255.1 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 558029 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 637621 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 107274 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 751811 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P109825 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2067226 | BC024482.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2067226 | NM_028809.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2067226 | AK008255.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P180108 | AK037305.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P180108 | BC024482.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P180108 | NM_028809.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P180108 | AK008255.1 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21151646 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21151639 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21151640 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21151642 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21151658 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21151644 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21151660 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21151666 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21151665 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21151668 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21151673 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21151645 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21151670 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21151662 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21151650 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21151664 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21151653 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21151654 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21151657 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P109825 | - | - |
| Whole Mouse Genome G4122A | A_51_P180108 | AK037305.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P180108 | BC024482.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P180108 | NM_028809.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P180108 | AK008255.1 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P128438 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P109825 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P01829882 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P01829885 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P21151652 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P21151650 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P21151651 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P21151656 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P21151655 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21151664 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21151630 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21151668 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21151658 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21151690 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21151589 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21151682 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21151638 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21151597 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21151581 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21151608 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21151645 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21151674 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21151705 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21151700 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21151627 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21151681 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21151702 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21151576 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21151587 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21151639 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21151600 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21151694 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21151635 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21151665 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21151657 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21151660 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21151662 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21151708 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21151706 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21151584 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21151686 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21151616 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21151620 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21151696 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21151634 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21151596 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21151583 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21151640 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21151692 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21151687 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21151654 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21151619 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21151585 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21151666 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21151594 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21151642 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21151680 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21151586 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21151606 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21151647 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21151624 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21151650 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21151598 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21151591 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21151688 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21151631 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21151617 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21151644 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21151579 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21151679 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21151704 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21151676 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21151650 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P02011056 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21151698 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P02087472 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21151666 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21151670 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21151668 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21151665 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21151702 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01897105 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21151662 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21151687 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P02237922 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21151654 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21151706 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21151699 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21151690 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01829870 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21151657 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21151664 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P02237918 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21151700 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01193571 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21151684 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21151708 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21151676 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21151681 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21151696 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01779856 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P02237921 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21151645 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21151694 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21151644 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21151646 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21151704 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21151688 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21151692 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21151658 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01897114 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21151647 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10471905 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10471906 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10471908 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10471907 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1441190_PM_at | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1444011_PM_at | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1452044_PM_at | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1436421_PM_s_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10471904 | - | - |
| Mouse Genome 430 2.0 Array | 1441190_PM_at | - | - |
| Mouse Genome 430 2.0 Array | 1444011_PM_at | - | - |
| Mouse Genome 430 2.0 Array | 1446087_at | - | - |
| Mouse Genome 430 2.0 Array | 1441190_at | - | - |
| Mouse Genome 430 2.0 Array | 1452044_at | - | - |
| Mouse Genome 430 2.0 Array | 1444011_at | - | - |
| Mouse Genome 430 2.0 Array | 1436421_PM_s_at | - | - |
| Mouse Genome 430 2.0 Array | 1436421_s_at | - | - |
| Mouse Genome 430 2.0 Array | 1446087_PM_at | - | - |
| Mouse Genome 430 2.0 Array | 1452044_PM_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 74192_PM_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 74192_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR0200006320.mm.1 | NM_028809.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0200006320.mm.1 | NM_028809.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0200006320.mm.1 | TC0200000719.mm.1 | MTA TC |
| Mouse Expression Set 430 | 1452044_PM_at | - | - |
| Mouse Expression Set 430 | 1452044_at | - | - |
| Mouse Expression Set 430 | 1436421_s_at | - | - |
| Mouse Expression Set 430 | 1436421_PM_s_at | - | - |
| Mouse Expression Set 430 | 1444011_at | - | - |
| Mouse Expression Set 430 | 1441190_PM_at | - | - |
| Mouse Expression Set 430 | 1441190_at | - | - |
| Mouse Expression Set 430 | 1444011_PM_at | - | - |
| Murine Genome U74v2 Set | 95538_PM_at | - | - |
| Murine Genome U74v2 Set | 95538_at | - | - |
| Murine Genome U74v2 Set | 161838_f_at | - | - |
| Murine Genome U74v2 Set | 161838_PM_f_at | - | - |
| Murine Genome U74v2 Set | 166018_PM_r_at | - | - |
| Murine Genome U74v2 Set | 165760_PM_f_at | - | - |
| Murine Genome U74v2 Set | 165760_f_at | - | - |
| Murine Genome U74v2 Set | 166018_r_at | - | - |
| Mouse Genome 430A 2.0 Array | 1452044_at | - | - |
| Mouse Genome 430A 2.0 Array | 1436421_PM_s_at | - | - |
| Mouse Genome 430A 2.0 Array | 1452044_PM_at | - | - |
| Mouse Genome 430A 2.0 Array | 1436421_s_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1436421_s_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1452044_PM_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1452044_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1436421_PM_s_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1441190_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1444011_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1444011_PM_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1441190_PM_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 744620 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina mouse-6 v1.0 expression beadchip | 1090148 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 104760010 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0001090148 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2626260 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_2626260 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_1249677 | - | - |
| mouse-6 v1.1 (Illumina) | scl20983.4_177-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 104760010 | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 1090148 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl20983.4_177-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 104760010 | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 1090148 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_2626260 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_1249677 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2626260 | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (3) | scl20983.4_177-S | - | - |

Arpc5l
Genomic Map