Assay Details
| Gene Symbol: Pip4k2a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_008845.4 | NP_032871.3 | 8 - 9 | 1164 | 140 | |
| GenBank mRNA | AB009615.1 | - | 8 - 9 | 1089 | 140 | |
| AK012196.1 | - | 8 - 9 | 1167 | 140 | ||
| AK040558.1 | - | 8 - 9 | 1054 | 140 | ||
| BC011097.1 | - | 8 - 9 | 1111 | 3672732 | 140 | |
More Information
Updated Mapping Information
Set Membership: |
|

Pip4k2a
Genomic Map