Assay Details
| Gene Symbol: Ushbp1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_181418.3 | NP_852083.2 | 10 - 11 | 1821 | 96 | |
| GenBank mRNA | AK043798.1 | - | 10 - 11 | 1759 | 96 | |
| AK084049.1 | - | 7 - 8 | 1268 | 96 | ||
| BC026453.1 | - | 10 - 11 | 1805 | 4460920 | 96 | |
More Information
Updated Mapping Information
Set Membership: |
|

Ushbp1
Genomic Map