Assay Details
| Gene Symbol: Ushbp1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_181418.3 | NP_852083.2 | 11 - 12 | 1909 | 101 | |
| GenBank mRNA | AK043798.1 | - | 11 - 12 | 1847 | 101 | |
| AK084049.1 | - | 8 - 9 | 1356 | 101 | ||
| BC026453.1 | - | 11 - 12 | 1893 | 4460920 | 101 | |
| BC033912.1 | - | 5 - 6 | 1498 | 101 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Ushbp1
Genomic Map