Assay Details
| Gene Symbol: Mak | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001145802.1 | NP_001139274.1 | 9 - 10 | 1553 | 135 | |
| NM_001145803.1 | NP_001139275.1 | 10 - 11 | 1646 | 135 | ||
| NM_008547.2 | NP_032573.2 | 9 - 10 | 1541 | 135 | ||
| GenBank mRNA | BC050009.1 | - | 9 - 10 | 5416 | 135 | |
| X66983.1 | - | 9 - 10 | 1535 | 135 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Mak
Genomic Map