Assay Details
| Gene Symbol: Mak | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001145803.1 | NP_001139275.1 | 4 - 5 | 610 | 167 | |
| NM_008547.2 | NP_032573.2 | 3 - 4 | 505 | 167 | ||
| GenBank mRNA | BC050009.1 | - | 4 - 5 | 549 | 167 | |
| BQ930470.1 | - | 4 - 5 | 525 | 167 | ||
| X66983.1 | - | 3 - 4 | 499 | 167 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Mak
Genomic Map