Assay Details
| Gene Symbol: Fkbp9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_012056.2 | NP_036186.2 | 5 - 6 | 1043 | 123 | |
| GenBank mRNA | AF090334.1 | - | 5 - 6 | 969 | 123 | |
| AF279263.1 | - | 5 - 6 | 1021 | 123 | ||
| AK007499.1 | - | 3 - 4 | 433 | 123 | ||
| BC026133.1 | - | 5 - 6 | 983 | 4018235 | 123 | |
| BC043129.1 | - | 5 - 6 | 1043 | 6810029 | 123 | |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Fkbp9
Genomic Map