Assay Details
| Gene Symbol: Fkbp9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_012056.2 | NP_036186.2 | 6 - 7 | 1192 | 76 | |
| GenBank mRNA | AF090334.1 | - | 6 - 7 | 1118 | 76 | |
| AK007499.1 | - | 4 - 5 | 582 | 76 | ||
| BC043129.1 | - | 6 - 7 | 1192 | 6810029 | 76 | |
More Information
Set Membership: |
|

Fkbp9
Genomic Map