Assay Details
| Gene Symbol: Tfe3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001105196.1 | NP_001098666.1 | 3 - 4 | 820 | 93 | |
| NM_001105197.1 | NP_001098667.1 | 3 - 4 | 506 | 93 | ||
| NM_001271489.1 | NP_001258418.1 | 3 - 4 | 801 | 93 | ||
| NM_001271490.1 | NP_001258419.1 | 3 - 4 | 973 | 93 | ||
| NM_001271491.1 | NP_001258420.1 | 3 - 4 | 540 | 93 | ||
| NM_172472.3 | NP_766060.2 | 3 - 4 | 820 | 93 | ||
| GenBank mRNA | AK089762.1 | - | 2 - 3 | 582 | 93 | |
| AK139756.1 | - | 3 - 4 | 817 | 93 | ||
| AK144657.1 | - | 3 - 4 | 785 | 93 | ||
| AK144751.1 | - | 3 - 4 | 788 | 93 | ||
| AK145029.1 | - | 3 - 4 | 940 | 93 | ||
| AK147405.1 | - | 2 - 3 | 1275 | 93 | ||
| AK150372.1 | - | 3 - 4 | 572 | 93 | ||
| AK151447.1 | - | 3 - 4 | 491 | 93 | ||
| AK152221.1 | - | 3 - 4 | 491 | 93 | ||
| AK153051.1 | - | 3 - 4 | 572 | 93 | ||
| AK155883.1 | - | 3 - 4 | 542 | 93 | ||
| AK159162.1 | - | 3 - 4 | 701 | 93 | ||
| BC056358.1 | - | 3 - 4 | 808 | 6813486 | 93 | |
| BC063047.1 | - | 3 - 4 | 808 | 6813486 | 93 | |
| S76673.1 | - | 1 - 2 | 158 | 93 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Tfe3
Genomic Map