Assay Details
| Gene Symbol: Lars2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_153168.2 | NP_694808.1 | 19 - 20 | 2586 | 67 | |
| GenBank mRNA | AJ428066.1 | - | 19 - 20 | 2586 | 67 | |
| AK032695.1 | - | 19 - 20 | 2591 | 67 | ||
| AK037012.1 | - | 19 - 20 | 2595 | 67 | ||
| AK053667.1 | - | 19 - 20 | 2581 | 67 | ||
| BC053916.1 | - | 20 - 21 | 2462 | 67 | ||
| BC080303.1 | - | 19 - 20 | 2473 | 5695617 | 67 | |
| BC112412.1 | - | 19 - 20 | 2555 | 40060997 | 67 | |
| BC138206.1 | - | 19 - 20 | 2560 | 8861225 | 67 | |
| BC145982.1 | - | 19 - 20 | 2560 | 40131285 | 67 | |
More Information
Set Membership: |
|

Lars2
Genomic Map