Assay Details
| Gene Symbol: Zdhhc3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_026917.5 | NP_081193.2 | 4 - 5 | 765 | 69 | |
| GenBank mRNA | AB088044.1 | - | 3 - 4 | 531 | 69 | |
| AK007355.1 | - | 5 - 6 | 1237 | 69 | ||
| AK018755.1 | - | 4 - 5 | 731 | 69 | ||
| AK080541.1 | - | 4 - 5 | 731 | 69 | ||
| AK169197.1 | - | 712 | 69 | |||
| AY166672.1 | - | 3 - 4 | 531 | 69 | ||
| BC005531.1 | - | 5 - 6 | 792 | 69 | ||
| BC018334.1 | - | 3 - 4 | 601 | 69 | ||
| BC025126.1 | - | 4 - 5 | 750 | 3669723 | 69 | |
| BC052464.1 | - | 4 - 5 | 670 | 6834763 | 69 | |
More Information
Updated Mapping Information
Set Membership: |
|

Zdhhc3
Genomic Map