Assay Details
| Gene Symbol: Dck | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_007832.4 | NP_031858.1 | 3 - 4 | 646 | 105 | |
| GenBank mRNA | AK004592.1 | - | 3 - 4 | 471 | 105 | |
| AK010700.1 | - | 3 - 4 | 436 | 105 | ||
| AK048347.1 | - | 3 - 4 | 483 | 105 | ||
| AK088195.1 | - | 3 - 4 | 483 | 105 | ||
| AK145353.1 | - | 4 - 5 | 592 | 105 | ||
| AK145390.1 | - | 4 - 5 | 592 | 105 | ||
| AK151851.1 | - | 3 - 4 | 480 | 105 | ||
| BC052004.1 | - | 3 - 4 | 570 | 6401517 | 105 | |
| BC060062.1 | - | 3 - 4 | 632 | 5703508 | 105 | |
| X77731.1 | - | 3 - 4 | 415 | 105 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Dck
Genomic Map