Assay Details
| Gene Symbol: Dck | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_007832.4 | NP_031858.1 | 5 - 6 | 902 | 69 | |
| GenBank mRNA | AK004592.1 | - | 5 - 6 | 727 | 69 | |
| AK010700.1 | - | 5 - 6 | 692 | 69 | ||
| AK048347.1 | - | 5 - 6 | 739 | 69 | ||
| AK088195.1 | - | 5 - 6 | 739 | 69 | ||
| AK145353.1 | - | 6 - 7 | 848 | 69 | ||
| AK145390.1 | - | 6 - 7 | 848 | 69 | ||
| AK151851.1 | - | 5 - 6 | 736 | 69 | ||
| BC052004.1 | - | 5 - 6 | 826 | 6401517 | 69 | |
| BC060062.1 | - | 5 - 6 | 888 | 5703508 | 69 | |
| X77731.1 | - | 5 - 6 | 671 | 69 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Dck
Genomic Map