Assay Details
| Gene Symbol: Tbc1d2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK148254.1 | - | 5 - 6 | 1156 | 69 | |
| AK148289.1 | - | 5 - 6 | 1070 | 69 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

Tbc1d2| Gene Symbol: Tbc1d2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK148254.1 | - | 5 - 6 | 1156 | 69 | |
| AK148289.1 | - | 5 - 6 | 1070 | 69 | ||
Set Membership: |
|