| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Mouse Clariom S Array |
PSR0400021261.mm.1 |
NM_198664.3.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0400021261.mm.1 |
NM_198664.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0400021261.mm.1 |
TC0400002605.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0400021263.mm.1 |
NM_198664.3.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0400021263.mm.1 |
NM_198664.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0400021263.mm.1 |
TC0400002605.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0400021268.mm.1 |
NM_198664.3.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0400021268.mm.1 |
NM_198664.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0400021268.mm.1 |
TC0400002605.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0400021262.mm.1 |
NM_198664.3.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0400021262.mm.1 |
NM_198664.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0400021262.mm.1 |
TC0400002605.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0400021264.mm.1 |
NM_198664.3.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0400021264.mm.1 |
NM_198664.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0400021264.mm.1 |
TC0400002605.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0400021269.mm.1 |
NM_198664.3.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0400021269.mm.1 |
NM_198664.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0400021269.mm.1 |
TC0400002605.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0400021260.mm.1 |
NM_198664.3.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0400021260.mm.1 |
NM_198664.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0400021260.mm.1 |
TC0400002605.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0400021265.mm.1 |
NM_198664.3.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0400021265.mm.1 |
NM_198664.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0400021265.mm.1 |
TC0400002605.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0400021267.mm.1 |
NM_198664.3.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0400021267.mm.1 |
NM_198664.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0400021267.mm.1 |
TC0400002605.mm.1 |
MTA TC |