Assay Details
| Gene Symbol: Brd9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK142152.1 | - | 5 - 6 | 663 | 97 | |
| AK154645.1 | - | 3 - 4 | 2264 | 97 | ||
| AK163238.1 | - | 3 - 4 | 2248 | 97 | ||
| BC031484.1 | - | 5 - 6 | 658 | 2654265 | 97 | |
| BC047372.1 | - | 5 - 6 | 642 | 97 | ||
| BC057597.1 | - | 3 - 4 | 2226 | 97 | ||
| BC089379.1 | - | 5 - 6 | 616 | 97 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Brd9
Genomic Map