Assay Details
| Gene Symbol: Hrasls | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK004528.1 | - | 1 - 2 | 155 | 65 | |
| BC048482.1 | - | 1 - 2 | 125 | 6774856 | 65 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the AK004528.1, BC048482.1 transcripts.
Set Membership: |
|

Hrasls
Genomic Map