Assay Details
| Gene Symbol: B4galt4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001285793.1 | NP_001272722.1 | 5 - 6 | 1120 | 64 | |
| NM_019804.4 | NP_062778.2 | 5 - 6 | 1053 | 64 | ||
| GenBank mRNA | AF142672.1 | - | 5 - 6 | 1030 | 64 | |
| AF158746.1 | - | 5 - 6 | 1032 | 64 | ||
| AK041692.1 | - | 5 - 6 | 1045 | 64 | ||
| AK045602.1 | - | 5 - 6 | 1025 | 64 | ||
| AK050005.1 | - | 5 - 6 | 1049 | 64 | ||
| AK052013.1 | - | 5 - 6 | 1047 | 64 | ||
| AK053603.1 | - | 5 - 6 | 1114 | 64 | ||
| AK085368.1 | - | 5 - 6 | 1039 | 64 | ||
| BC013492.1 | - | 5 - 6 | 1016 | 4218187 | 64 | |
| BC031115.1 | - | 5 - 6 | 1105 | 5026168 | 64 | |
More Information
Updated Mapping Information
Set Membership: |
|

B4galt4
Genomic Map