| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Mouse Clariom S Array |
PSR1300004372.mm.1 |
NM_001038018.4 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1300004372.mm.1 |
NM_001112711.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1300004372.mm.1 |
NM_001286063.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1300004372.mm.1 |
NM_001286064.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1300004372.mm.1 |
NM_001286065.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1300004372.mm.1 |
NM_001286066.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1300004372.mm.1 |
NM_011938.4 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1300004372.mm.1 |
TC1300000660.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1300004371.mm.1 |
NM_001038018.4 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1300004371.mm.1 |
NM_001112711.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1300004371.mm.1 |
NM_001286063.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1300004371.mm.1 |
NM_001286064.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1300004371.mm.1 |
NM_001286065.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1300004371.mm.1 |
NM_001286066.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1300004371.mm.1 |
NM_011938.4 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1300004371.mm.1 |
TC1300000660.mm.1 |
MTA TC |