Assay Details
| Gene Symbol: Scn5a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001253860.1 | NP_001240789.1 | 3 - 4 | 533 | 103 | |
| NM_021544.4 | NP_067519.2 | 4 - 5 | 701 | 103 | ||
| GenBank mRNA | AJ271477.1 | - | 3 - 4 | 529 | 103 | |
| AK147254.1 | - | 4 - 5 | 701 | 103 | ||
| AK147517.1 | - | 4 - 5 | 701 | 103 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Scn5a
Genomic Map