Assay Details
| Gene Symbol: Lsg1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_178069.5 | NP_835170.1 | 7 - 8 | 811 | 91 | |
| GenBank mRNA | AK030940.1 | - | 7 - 8 | 786 | 91 | |
| AK078501.1 | - | 7 - 8 | 784 | 91 | ||
| AK132527.1 | - | 8 - 9 | 1014 | 91 | ||
| AK135345.1 | - | 806 | 91 | |||
| AK146895.1 | - | 7 - 8 | 786 | 91 | ||
| BC043724.1 | - | 7 - 8 | 811 | 5004563 | 91 | |
| BC080306.1 | - | 5 - 6 | 542 | 91 | ||
More Information
Set Membership: |
|

Lsg1
Genomic Map