Assay Details
| Gene Symbol: Lsg1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_178069.5 | NP_835170.1 | 8 - 9 | 1187 | 127 | |
| GenBank mRNA | AK030940.1 | - | 8 - 9 | 1162 | 127 | |
| AK078501.1 | - | 8 - 9 | 1160 | 127 | ||
| AK132527.1 | - | 9 - 10 | 1390 | 127 | ||
| AK135345.1 | - | 1182 | 127 | |||
| AK146895.1 | - | 8 - 9 | 1162 | 127 | ||
| BC043724.1 | - | 8 - 9 | 1187 | 5004563 | 127 | |
| BC080306.1 | - | 6 - 7 | 918 | 127 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Lsg1
Genomic Map