Assay Details
| Gene Symbol: Lsg1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_178069.5 | NP_835170.1 | 9 - 10 | 1282 | 83 | |
| GenBank mRNA | AK030940.1 | - | 9 - 10 | 1257 | 83 | |
| AK078501.1 | - | 9 - 10 | 1255 | 83 | ||
| AK132527.1 | - | 10 - 11 | 1485 | 83 | ||
| AK135345.1 | - | 1277 | 83 | |||
| AK145186.1 | - | 9 - 10 | 1260 | 83 | ||
| AK146895.1 | - | 9 - 10 | 1257 | 83 | ||
| BC043724.1 | - | 9 - 10 | 1282 | 5004563 | 83 | |
| BC080306.1 | - | 7 - 8 | 1013 | 83 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Lsg1
Genomic Map