Assay Details
| Gene Symbol: Tmem44 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_172614.3 | NP_766202.3 | 7 - 8 | 1004 | 117 | |
| GenBank mRNA | AK031524.1 | - | 6 - 7 | 3456 | 117 | |
| AK045675.1 | - | 8 - 9 | 1067 | 117 | ||
| AK048655.1 | - | 7 - 8 | 1005 | 117 | ||
| AK079084.1 | - | 2 - 3 | 330 | 117 | ||
| AK082652.1 | - | 7 - 8 | 2239 | 118 | ||
| BC059216.1 | - | 7 - 8 | 1342 | 117 | ||
| BC080307.1 | - | 8 - 9 | 900 | 117 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Tmem44
Genomic Map