Assay Details
| Gene Symbol: Mcm4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_008565.3 | NP_032591.3 | 3 - 4 | 616 | 64 | |
| GenBank mRNA | AK011743.1 | - | 3 - 4 | 551 | 64 | |
| AK036653.1 | - | 3 - 4 | 576 | 64 | ||
| AK083618.1 | - | 3 - 4 | 530 | 64 | ||
| AK088796.1 | - | 3 - 4 | 536 | 64 | ||
| AK089999.1 | - | 3 - 4 | 520 | 64 | ||
| AK135422.1 | - | 3 - 4 | 518 | 64 | ||
| AK141132.1 | - | 3 - 4 | 582 | 64 | ||
| AK141644.1 | - | 3 - 4 | 617 | 64 | ||
| AK146662.1 | - | 3 - 4 | 539 | 64 | ||
| AK151499.1 | - | 3 - 4 | 530 | 64 | ||
| AK157725.1 | - | 3 - 4 | 518 | 64 | ||
| AK220442.1 | - | 3 - 4 | 517 | 64 | ||
| BC013094.1 | - | 3 - 4 | 572 | 3584640 | 64 | |
| D26089.1 | - | 3 - 4 | 522 | 64 | ||
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Mcm4
Genomic Map