Assay Details
| Gene Symbol: Cblb | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001033238.1 | NP_001028410.1 | 6 - 7 | 999 | 62 | |
| GenBank mRNA | AK156295.1 | - | 7 - 8 | 1030 | 62 | |
| AK161486.1 | - | 4 - 5 | 585 | 62 | ||
| AK170640.1 | - | 6 - 7 | 943 | 62 | ||
| BC150934.1 | - | 6 - 7 | 955 | 9087845 | 62 | |
| BC150938.1 | - | 6 - 7 | 955 | 9087849 | 62 | |
More Information
Updated Mapping Information
Set Membership: |
|

Cblb
Genomic Map