Assay Details
| Gene Symbol: Zpld1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_178720.4 | NP_848835.1 | 6 - 7 | 737 | 152 | |
| GenBank mRNA | AK034628.1 | - | 6 - 7 | 738 | 152 | |
| AK047981.1 | - | 5 - 6 | 647 | 152 | ||
| AK052599.1 | - | 7 - 8 | 847 | 152 | ||
| BC116254.1 | - | 6 - 7 | 706 | 40093631 | 152 | |
| BC116255.1 | - | 6 - 7 | 706 | 40093632 | 152 | |
More Information
Updated Mapping Information
Set Membership: |
|

Zpld1
Genomic Map