Assay Details
| Gene Symbol: Map3k13 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_172821.3 | NP_766409.2 | 4 - 5 | 1010 | 59 | |
| GenBank mRNA | DQ480427.1 | - | 4 - 5 | 1010 | 59 | |
More Information
Updated Mapping Information
Set Membership: |
|

Map3k13
Genomic Map