Assay Details
| Gene Symbol: Zbtb20 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001285805.1 | NP_001272734.1 | 1 - 2 | 80 | 80 | |
| GenBank mRNA | AK132682.1 | - | 2 - 3 | 81 | 80 | |
| BC056446.1 | - | 1 - 2 | 52 | 5703319 | 80 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the BC056446.1, NM_001285805.1 transcripts.
Set Membership: |
|

Zbtb20
Genomic Map