Assay Details
| Gene Symbol: Gbe1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_028803.4 | NP_083079.1 | 11 - 12 | 1647 | 81 | |
| GenBank mRNA | AK009815.1 | - | 11 - 12 | 1463 | 81 | |
| AK050365.1 | - | 11 - 12 | 1534 | 81 | ||
| AK050423.1 | - | 11 - 12 | 1533 | 81 | ||
| AK148186.1 | - | 5 - 6 | 575 | 81 | ||
| BC017541.1 | - | 11 - 12 | 1508 | 4526961 | 81 | |
More Information
Set Membership: |
|

Gbe1
Genomic Map