Assay Details
| Gene Symbol: Gbe1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_028803.4 | NP_083079.1 | 5 - 6 | 893 | 129 | |
| GenBank mRNA | AK009815.1 | - | 5 - 6 | 709 | 129 | |
| AK050365.1 | - | 5 - 6 | 780 | 129 | ||
| AK050423.1 | - | 5 - 6 | 779 | 129 | ||
| BC017541.1 | - | 5 - 6 | 754 | 4526961 | 129 | |
More Information
Updated Mapping Information
Set Membership: |
|

Gbe1
Genomic Map