Assay Details
| Gene Symbol: Gbe1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_028803.4 | NP_083079.1 | 8 - 9 | 1312 | 141 | |
| GenBank mRNA | AK009815.1 | - | 8 - 9 | 1128 | 141 | |
| AK050365.1 | - | 8 - 9 | 1199 | 141 | ||
| AK050423.1 | - | 8 - 9 | 1198 | 141 | ||
| AK148186.1 | - | 2 - 3 | 240 | 141 | ||
| BC017541.1 | - | 8 - 9 | 1173 | 4526961 | 141 | |
More Information
Set Membership: |
|

Gbe1
Genomic Map