Assay Details
| Gene Symbol: Rbm11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_198302.2 | NP_938044.1 | 1 - 2 | 149 | 88 | |
| GenBank mRNA | AK039297.1 | - | 1 - 2 | 91 | 88 | |
| BC050779.1 | - | 1 - 2 | 196 | 6771154 | 88 | |
More Information
Updated Mapping Information
Set Membership: |
|

Rbm11
Genomic Map